ENA405193966 Enamine

methyl(1H-1,2,3,4-tetrazol-5-ylmethyl)amine hydrochloride

  • MDL number MFCD25970429
  • Cas Number : 1461714-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239...241
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134300

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134300
Availability details for
Quantity
  • Availability
  • Add to Cart