ENA405193968 Enamine

1-(4-methoxyphenyl)-3-(1H-1,2,4-triazol-1-yl)propan-1-one

  • MDL number MFCD10574946
  • Cas Number : 108664-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68...70
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134312

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134312
Availability details for
Quantity
  • Availability
  • Add to Cart