ENA405193969 Enamine

ethyl 3-(2-chloro-6-nitrophenyl)-3-oxopropanoate

  • MDL number MFCD25970433
  • Cas Number : 1461705-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48...50
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134368

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134368
Availability details for
Quantity
  • Availability
  • Add to Cart