ENA405193971 Enamine

2-[(4-chlorophenyl)sulfanyl]-5-methylbenzonitrile

  • MDL number MFCD25970435
  • Cas Number : 1461714-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134378
Availability details for
Quantity
  • Availability
  • Add to Cart