ENA405193974 Enamine

4-[(cyclopropylamino)methyl]benzonitrile hydrochloride

  • MDL number MFCD25970438
  • Cas Number : 1461705-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134468

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134468
Availability details for
Quantity
  • Availability
  • Add to Cart