ENA405193976 Enamine

5-ethoxy-4-methyl-1,3-oxazole-2-carbohydrazide

  • MDL number MFCD25970440
  • Cas Number : 1461714-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134496
Availability details for
Quantity
  • Availability
  • Add to Cart