ENA405193979 Enamine

N-[2-(1-methyl-1H-1,3-benzodiazol-2-yl)propan-2-yl]benzamide

  • MDL number MFCD25970444
  • Cas Number : 1461714-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134524
Availability details for
Quantity
  • Availability
  • Add to Cart