ENA405193980 Enamine

4,6-difluoro-2,3-dihydro-1-benzofuran-3-amine hydrochloride

  • MDL number MFCD25970445
  • Cas Number : 1461713-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 243...245
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134533

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134533
Availability details for
Quantity
  • Availability
  • Add to Cart