ENA405193981 Enamine

2-ethyl-1,3-thiazinane-4-carboxylic acid hydrochloride

  • MDL number MFCD25970447
  • Cas Number : 1461706-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203...205
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134558

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134558
Availability details for
Quantity
  • Availability
  • Add to Cart