ENA405193983 Enamine

3-(aminomethyl)-4,4-dimethylpentanoic acid hydrochloride

  • MDL number MFCD25970449
  • Cas Number : 1461713-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134560

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134560
Availability details for
Quantity
  • Availability
  • Add to Cart