ENA405193985 Enamine

1-[4-(aminomethyl)phenyl]ethan-1-amine dihydrochloride

  • MDL number MFCD25970452
  • Cas Number : 1461714-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 299...301
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134565

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134565
Availability details for
Quantity
  • Availability
  • Add to Cart