ENA405193987 Enamine

4,5,5-trimethylpiperidin-2-one

  • MDL number MFCD25970456
  • Cas Number : 1461704-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134592
Availability details for
Quantity
  • Availability
  • Add to Cart