ENA405193989 Enamine

3-[ethyl(2,2,2-trifluoroethyl)amino]propan-1-ol hydrochloride

  • MDL number MFCD25970459
  • Cas Number : 1461714-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134642

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134642
Availability details for
Quantity
  • Availability
  • Add to Cart