ENA405193991 Enamine

ethyl 2-[1-(chlorosulfonyl)cyclopropyl]acetate

  • MDL number MFCD25970462
  • Cas Number : 1461708-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134682
Availability details for
Quantity
  • Availability
  • Add to Cart