ENA405193995 Enamine

3-(2-aminoacetamido)-N-[(pyridin-2-yl)methyl]propanamide dihydrochloride

  • MDL number MFCD25970467
  • Cas Number : 1461713-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134698
Availability details for
Quantity
  • Availability
  • Add to Cart