ENA405194000 Enamine

methyl[2-(1H-1,2,3,4-tetrazol-5-yl)propyl]amine hydrochloride

  • MDL number MFCD25970473
  • Cas Number : 1461708-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134838

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134838
Availability details for
Quantity
  • Availability
  • Add to Cart