ENA405194009 Enamine

(1R)-1-cycloheptylethan-1-amine hydrochloride

  • MDL number MFCD25970488
  • Cas Number : 1461689-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229-231
Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134923
Availability details for
Quantity
  • Availability
  • Add to Cart