ENA405194011 Enamine

2-(oxan-2-yl)ethyl methanesulfonate

  • MDL number MFCD25970490
  • Cas Number : 133243-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134981
Availability details for
Quantity
  • Availability
  • Add to Cart