ENA405194019 Enamine

(2S)-2-amino-3-cycloheptylpropanoic acid hydrochloride

  • MDL number MFCD25970498
  • Cas Number : 1461689-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135144
Availability details for
Quantity
  • Availability
  • Add to Cart