ENA405194021 Enamine

1-(2,2-difluoroethyl)piperidin-4-amine dihydrochloride

  • MDL number MFCD25970500
  • Cas Number : 1201694-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135178
Availability details for
Quantity
  • Availability
  • Add to Cart