ENA405194023 Enamine

2-(5-methyl-1,2,4-oxadiazol-3-yl)azepane hydrochloride

  • MDL number MFCD25970502
  • Cas Number : 1461713-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135208
Availability details for
Quantity
  • Availability
  • Add to Cart