ENA405194028 Enamine

3-ethyl-5-methyl-1,2-oxazole-4-carbonitrile

  • MDL number MFCD25970289
  • Cas Number : 1461706-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-131166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131166
Availability details for
Quantity
  • Availability
  • Add to Cart