ENA405194029 Enamine

methyl 2-(2-acetylphenoxy)acetate

  • MDL number MFCD00119145
  • Cas Number : 77956-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131181
Availability details for
Quantity
  • Availability
  • Add to Cart