ENA405194030 Enamine

2,5-dimethylfuran-3-carbonitrile

  • MDL number MFCD12405644
  • Cas Number : 89149-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131183
Availability details for
Quantity
  • Availability
  • Add to Cart