ENA405194035 Enamine

3-(2-nitroethenyl)-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD21648942
  • Cas Number : 183208-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 244...246
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131357

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131357
Availability details for
Quantity
  • Availability
  • Add to Cart