ENA405194042 Enamine

N-phenylpiperidine-3-sulfonamide

  • MDL number MFCD09936263
  • Cas Number : 1016833-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131511
Availability details for
Quantity
  • Availability
  • Add to Cart