ENA405194056 Enamine

3-bromo-2-chloro-4-methyl-5-nitropyridine

  • MDL number MFCD12024452
  • Cas Number : 1049706-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62...64
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131745

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131745
Availability details for
Quantity
  • Availability
  • Add to Cart