ENA405194058 Enamine

4-nitro-2-(pyridin-2-yl)-1H-1,3-benzodiazole

  • MDL number MFCD25970298
  • Cas Number : 68118-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217...219
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131756

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131756
Availability details for
Quantity
  • Availability
  • Add to Cart