ENA405194060 Enamine

4-(propan-2-yl)morpholine-2-carbonitrile

  • MDL number MFCD25970256
  • Cas Number : 1461715-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100532

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100532
Availability details for
Quantity
  • Availability
  • Add to Cart