ENA405194069 Enamine

4-(cyclopentylsulfanyl)quinoline-3-carboxylic acid

  • MDL number MFCD19110275
  • Cas Number : 1275908-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132033
Availability details for
Quantity
  • Availability
  • Add to Cart