ENA405194070 Enamine

2-(chloromethyl)-4-cyclopropyl-1,3-thiazole

  • MDL number MFCD19606981
  • Cas Number : 170881-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132035
Availability details for
Quantity
  • Availability
  • Add to Cart