ENA405194075 Enamine

1,4-dimethylpiperidine-4-carboxylic acid hydrochloride

  • MDL number MFCD11841301
  • Cas Number : 1185325-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 246...248
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132136

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132136
Availability details for
Quantity
  • Availability
  • Add to Cart