ENA405194076 Enamine

5-methyl-1-(2-nitrophenyl)-1H-pyrazole

  • MDL number MFCD15527658
  • Cas Number : 1247439-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132150

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132150
Availability details for
Quantity
  • Availability
  • Add to Cart