ENA405194077 Enamine

1-cyclohexylcyclopropane-1-carboxylic acid

  • MDL number MFCD20657665
  • Cas Number : 803736-51-8
  • Molecular Weight : 168.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclohexylcyclopropanecarboxylic acid
Inchi Code 1S/C10H16O2/c11-9(12)10(6-7-10)8-4-2-1-3-5-8/h8H,1-7H2,(H,11,12)
Inchi Key CXFVLOHIQZJBGH-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-132177

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-132177
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart