ENA405194080 Enamine

methyl 2-[(3,4-difluorophenyl)(hydroxy)methyl]prop-2-enoate

  • MDL number MFCD11130338
  • Cas Number : 1019127-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132188
Availability details for
Quantity
  • Availability
  • Add to Cart