ENA405194081 Enamine

2-iodo-5-methoxypyridine

  • MDL number MFCD02259696
  • Cas Number : 163129-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132210

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132210
Availability details for
Quantity
  • Availability
  • Add to Cart