ENA405194083 Enamine

3-(2,5-difluorophenoxy)propanoic acid

  • MDL number MFCD18305843
  • Cas Number : 942485-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82...84
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132243

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132243
Availability details for
Quantity
  • Availability
  • Add to Cart