ENA405194084 Enamine

ethyl 2-(1-benzylpiperidin-3-ylidene)acetate

  • MDL number MFCD25970304
  • Cas Number : 150543-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132247

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132247
Availability details for
Quantity
  • Availability
  • Add to Cart