ENA405194092 Enamine

3,5-dimethyl-1-(2-methylpropyl)-1H-pyrazole

  • MDL number MFCD08701136
  • Cas Number : 79168-92-6
  • Molecular Weight : 152.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-132355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132355
Availability details for
Quantity
  • Availability
  • Add to Cart