ENA405194096 Enamine

2-(4-nitrobenzenesulfonyl)ethan-1-ol

  • MDL number MFCD23702250
  • Cas Number : 21386-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128...130
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132430

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132430
Availability details for
Quantity
  • Availability
  • Add to Cart