ENA405194097 Enamine

5-chloro-2-(cyclopentylsulfanyl)benzonitrile

  • MDL number MFCD18270313
  • Cas Number : 1407356-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132431

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132431
Availability details for
Quantity
  • Availability
  • Add to Cart