ENA405194098 Enamine

2-(butylsulfanyl)-5-chlorobenzonitrile

  • MDL number MFCD16769724
  • Cas Number : 1249077-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45...47
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132432

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132432
Availability details for
Quantity
  • Availability
  • Add to Cart