ENA405194129 Enamine

1-chloro-2-(3-chloropropyl)benzene

  • MDL number MFCD06656680
  • Cas Number : 90347-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132842

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132842
Availability details for
Quantity
  • Availability
  • Add to Cart