ENA405194130 Enamine

1-cyclopropyl-1-methylthiourea

  • MDL number MFCD11624020
  • Cas Number : 1094883-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132846

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132846
Availability details for
Quantity
  • Availability
  • Add to Cart