ENA405194138 Enamine

3-amino-1-(pyridin-3-yl)piperidin-2-one

  • MDL number MFCD19679343
  • Cas Number : 1339471-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132910

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132910
Availability details for
Quantity
  • Availability
  • Add to Cart