ENA405194140 Enamine

1,1,1-trichloro-3-(quinolin-2-yl)propan-2-ol

  • MDL number MFCD00030689
  • Cas Number : 56211-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132977
Availability details for
Quantity
  • Availability
  • Add to Cart