ENA405194142 Enamine

5-[3-(trifluoromethyl)phenyl]-1,3-oxazole-4-carbonitrile

  • MDL number MFCD00108300
  • Cas Number : 255876-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101...103
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132981

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132981
Availability details for
Quantity
  • Availability
  • Add to Cart