ENA405194146 Enamine

4-(benzyloxy)but-2-enoic acid

  • Cas Number : 162551-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133007
Availability details for
Quantity
  • Availability
  • Add to Cart