ENA405194152 Enamine

1-(4-fluoro-2-nitrophenyl)ethan-1-one

  • MDL number MFCD13194367
  • Cas Number : 1214346-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133067
Availability details for
Quantity
  • Availability
  • Add to Cart