ENA405194154 Enamine

4-(diethoxymethyl)-1-(4-nitrophenyl)-1H-1,2,3-triazole

  • MDL number MFCD25970318
  • Cas Number : 1012081-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70...72
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133076

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133076
Availability details for
Quantity
  • Availability
  • Add to Cart